Assay Details
Gene Symbol: CCDC154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143980.1 | NP_001137452.1 | 10 - 11 | 1315 | 103 | |
GenBank mRNA | JN935901.1 | - | 9 - 10 | 1148 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000752.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16010985.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16010985.hg.1 | ENST00000409671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16010985.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16010985.hg.1 | TC16000752.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16010984.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16010984.hg.1 | ENST00000409671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16010984.hg.1 | ENST00000483702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16010984.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16010984.hg.1 | TC16000752.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | TC1600009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168427.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | TC1600009041.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600168428.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000389176.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000409671.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000463299.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | ENST00000483702.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009041.hg.1 | NM_001143980.1 | Refseq |
