Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000422548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000422548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000422548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054252.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054252.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054252.hg.1 |
TC01003488.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000422548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000422548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000422548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016333.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224223.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224223.hg.1 |
NR_037851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224223.hg.1 |
TC0100016333.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100224223.hg.1 |
ENST00000441851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224223.hg.1 |
NR_037851.1 |
Refseq |