Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016429.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016429.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016429.hg.1 |
TC09001315.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016428.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016428.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016428.hg.1 |
TC09001315.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010723.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173035.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173035.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173035.hg.1 |
TC0900010723.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173035.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173035.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173036.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173036.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173036.hg.1 |
TC0900010723.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173036.hg.1 |
ENST00000426593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173036.hg.1 |
NR_038882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008241.hg.1 |
ENST00000514769.1 |
Ensembl |