Assay Details
Gene Symbol: MIR4290HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038882.1 | 2 - 3 | 293 | 69 | ||
GenBank mRNA | BC039331.1 | - | 2 - 3 | 293 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4290 | MIR4290 | -1.0kb | Sense | Chr. 9 - 90023441 - 90023535 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1508 | LINC01508 | 264.8kb | Sense | Chr. 9 - 90300896 - 90433489 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001315.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
