Assay Details
Gene Symbol: LOC101927865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BI830187.1 | - | 4 - 5 | 681 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
