Assay Details
Gene Symbol: LOC101927211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110742.1 | 1 - 2 | 139 | 138 | ||
GenBank mRNA | BC032020.1 | - | 1 - 2 | 139 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
