Assay Details
Gene Symbol: HNF4A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109949.1 | 2 - 3 | 352 | 77 | ||
GenBank mRNA | AF143870.1 | - | 2 - 3 | 363 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
