Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000455642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000455642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000609152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000609152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
ENST00000609152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164223.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164223.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164223.hg.1 |
TC2000009188.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164223.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164223.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000452481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000455642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000455642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000609152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000609152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
ENST00000609152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009188.hg.1 |
NR_109949.1 |
Refseq |