Assay Details
Gene Symbol: KANSL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034172.1 | 1 - 2 | 159 | 153 | ||
GenBank mRNA | BC020847.1 | - | 1 - 2 | 159 | 153 | |
BC066350.1 | - | 1 - 2 | 156 | 153 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000596.hg.1, TC17_ctg5_hap1000016.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10341623 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P221327 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 19987 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3220105 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P221327 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P221327 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 229857_PM_s_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 230388_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR17007274.hg.1 | ENST00000398275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007274.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007274.hg.1 | TC17000596.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17007275.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007275.hg.1 | TC17000596.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000211.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000211.hg.1 | TC17_ctg5_hap1000016.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007273.hg.1 | ENST00000398275.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007273.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007273.hg.1 | TC17000596.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000212.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17_ctg5_hap1000212.hg.1 | TC17_ctg5_hap1000016.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700159284.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159284.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159284.hg.1 | TC1700008091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159284.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159284.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | TC1700008091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159288.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008091.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | TC1700008091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | ENST00000572634.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159289.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | NR_034172.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | TC1700008091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | ENST00000398275.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | ENST00000572973.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700159285.hg.1 | NR_034172.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 229857_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 230388_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 230388_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 229857_PM_s_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 230388_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229857_s_at8 | - | - |
Human Genome U95 Set | 64461_PM_at | - | - |
Human Genome U95 Set | 64461_at | - | - |
Human Genome U133 Set | 230388_PM_s_at | - | - |
Human Genome U133 Set | 229857_s_at | - | - |
Human Genome U133 Set | 229857_PM_s_at | - | - |
Human Genome U133 Set | 230388_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009621 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009621 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 229949 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1755200 | - | - |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1755200 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1755200 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1755200 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1755200 | - | - |
