Assay Details
Gene Symbol: TTBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032538.1 | NP_115927.1 | 14 - 15 | 3850 | 127 | |
XM_017011365.1 | XP_016866854.1 | 12 - 13 | 3901 | 127 | ||
GenBank mRNA | AB058758.1 | - | 12 - 13 | 3419 | 127 | |
AB218664.1 | - | 15 - 16 | 3654 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000598.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007068.hg.1 | ENST00000259750.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007068.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007068.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007069.hg.1 | ENST00000259750.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007069.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007069.hg.1 | TC06000598.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | ENST00000304139.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008075.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156380.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156380.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156380.hg.1 | TC0600008075.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600156380.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156380.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156381.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156381.hg.1 | NM_032538.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600156381.hg.1 | TC0600008075.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600156381.hg.1 | ENST00000259750.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600156381.hg.1 | NM_032538.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 133658 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 637621 | - | - |
