Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052917.hg.1 |
TC01006412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052917.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052917.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052919.hg.1 |
TC01006412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052919.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052919.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052916.hg.1 |
TC01006412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052916.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052916.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052918.hg.1 |
TC01006412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052918.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052918.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243767.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243767.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243767.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243767.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243767.hg.1 |
TC0100018514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100243768.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243768.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243768.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243768.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243768.hg.1 |
TC0100018514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100243765.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243765.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243765.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243765.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243765.hg.1 |
TC0100018514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018514.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243764.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243764.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243764.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243764.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243764.hg.1 |
TC0100018514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100243766.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243766.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243766.hg.1 |
ENST00000368100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100243766.hg.1 |
NM_001013661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100243766.hg.1 |
TC0100018514.hg.1 |
Clariom D TC |