Assay Details
Gene Symbol: LOC101929625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109880.1 | 2 - 3 | 362 | 95 | ||
GenBank mRNA | DA089649.1 | - | 2 - 3 | 363 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | TC2000006909.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147301.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | TC2000006909.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000147303.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000621402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000622928.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000623256.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624367.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | ENST00000624385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006909.hg.1 | NR_109880.1 | Refseq |
