Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027640.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027640.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027640.hg.1 |
TC06002206.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027639.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027639.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027639.hg.1 |
TC06002206.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600193253.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193253.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193253.hg.1 |
TC0600013507.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193253.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193253.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193254.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193254.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600193254.hg.1 |
TC0600013507.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600193254.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600193254.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
ENST00000413845.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013507.hg.1 |
NR_038408.1 |
Refseq |