Assay Details
Gene Symbol: UST-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038408.1 | 2 - 3 | 184 | 93 | ||
GenBank mRNA | BC034817.1 | - | 2 - 3 | 184 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002206.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3335975 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P248098 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 34520 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23758 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3335975 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3335975 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P248098 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P248098 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P370356 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1557742_PM_a_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1557741_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06027638.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06027638.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027638.hg.1 | TC06002206.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027637.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06027637.hg.1 | TC06002206.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0600193252.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600193252.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600193252.hg.1 | TC0600013507.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600193252.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600193252.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | ENST00000413845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013507.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600193251.hg.1 | NR_038408.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600193251.hg.1 | TC0600013507.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600193251.hg.1 | NR_038408.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1557741_at | - | - |
Human Genome U133 Plus 2.0 Array | 1557742_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1557741_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1557742_a_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557741_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1557742_a_at8 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030676 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030675 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030676 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030675 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 233639 | - | - |
