Assay Details
Gene Symbol: LOC101929645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109948.1 | 2 - 3 | 499 | 164 | ||
GenBank mRNA | AL137617.1 | - | 2 - 3 | 499 | 164 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | lymphocyte-specific protein 1 pseudogene 3 | LSP1P3 | -138.5kb | Sense | Chr. 5 - 28926870 - 28927313 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101929660 | LOC101929660 | 73.2kb | Sense | Chr. 5 - 29143503 - 29172975 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
