Assay Details
Gene Symbol: LOC105378933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135034.1 | 1 - 2 | 128 | 119 | ||
GenBank mRNA | AA429672.1 | - | 1 - 2 | 124 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
