Assay Details
Gene Symbol: CTB-12O2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK001582.1 | - | 4 - 5 | 278 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | NR_109876.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | NR_109876.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009135.hg.1 | NR_109876.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000518431.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000523605.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | ENST00000524295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | NR_109876.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | NR_109876.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009135.hg.1 | NR_109876.1 | Refseq |
