Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000515843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000515843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000515843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000624134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000624134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
ENST00000624134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000503243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000515843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000515843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000515843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000624134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000624134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
ENST00000624134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011703.hg.1 |
NR_125931.1 |
Refseq |