Assay Details
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098483.2 | NP_001091953.1 | 1 - 2 | 108 | 115 | |
NM_198472.2 | NP_940874.2 | 1 - 2 | 108 | 115 | ||
XM_011539670.2 | XP_011537972.1 | 1 - 2 | 114 | 115 | ||
XM_011539671.2 | XP_011537973.1 | 1 - 2 | 114 | 115 | ||
XM_011539672.2 | XP_011537974.1 | 1 - 2 | 114 | 115 | ||
XM_011539673.2 | XP_011537975.1 | 1 - 2 | 114 | 115 | ||
XM_011539674.2 | XP_011537976.1 | 1 - 2 | 114 | 115 | ||
XM_011539675.2 | XP_011537977.1 | 1 - 2 | 114 | 115 | ||
GenBank mRNA | AK129527.1 | - | 1 - 2 | 108 | 115 | |
BC129818.1 | - | 1 - 2 | 108 | 115 | ||
BC129819.1 | - | 1 - 2 | 108 | 40113019 | 115 | |
HY163253.1 | - | 1 - 2 | 103 | 115 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001789.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | ENST00000278025.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | ENST00000368552.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | ENST00000447176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | ENST00000465384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023633.hg.1 | TC10001789.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | ENST00000278025.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | ENST00000368552.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | ENST00000465384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023630.hg.1 | TC10001789.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | TC1000012398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184637.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
