Assay Details
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098483.2 | NP_001091953.1 | 3 - 4 | 247 | 73 | |
NM_001301827.1 | NP_001288756.1 | 2 - 3 | 178 | 73 | ||
NM_001301828.1 | NP_001288757.1 | 3 - 4 | 454 | 73 | ||
NM_198472.2 | NP_940874.2 | 3 - 4 | 247 | 73 | ||
XM_011539670.2 | XP_011537972.1 | 3 - 4 | 253 | 73 | ||
XM_011539671.2 | XP_011537973.1 | 3 - 4 | 253 | 73 | ||
XM_011539672.2 | XP_011537974.1 | 3 - 4 | 253 | 73 | ||
XM_011539673.2 | XP_011537975.1 | 3 - 4 | 253 | 73 | ||
XM_011539674.2 | XP_011537976.1 | 3 - 4 | 253 | 73 | ||
XM_011539675.2 | XP_011537977.1 | 3 - 4 | 253 | 73 | ||
XM_017016094.1 | XP_016871583.1 | 3 - 4 | 451 | 73 | ||
GenBank mRNA | AK129527.1 | - | 3 - 4 | 247 | 73 | |
BC129818.1 | - | 3 - 4 | 247 | 73 | ||
BC129819.1 | - | 3 - 4 | 247 | 40113019 | 73 | |
BM009909.1 | - | 2 - 3 | 110 | 73 | ||
BU849497.1 | - | 3 - 4 | 430 | 73 | ||
BX119427.1 | - | 3 - 4 | 173 | 73 | ||
HY163253.1 | - | 3 - 4 | 242 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
