Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003506.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003506.hg.1 |
ENST00000538876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003506.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003506.hg.1 |
TC0X000269.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000490300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
ENST00000496651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007224.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149574.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149574.hg.1 |
NM_014008.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149574.hg.1 |
TC0X00007224.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00149574.hg.1 |
ENST00000376227.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149574.hg.1 |
NM_014008.4 |
Refseq |