Assay Details
Gene Symbol: MAP3K10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002446.3 | NP_002437.2 | 2 - 3 | 1152 | 96 | |
XM_011526981.2 | XP_011525283.1 | 2 - 3 | 1239 | 96 | ||
XM_011526982.2 | XP_011525284.1 | 2 - 3 | 1238 | 96 | ||
GenBank mRNA | X90846.1 | - | 2 - 3 | 1152 | 96 | |
Z48615.1 | - | 2 - 3 | 1024 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000559.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction
smoothened signaling pathway
JNK cascade
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of apoptotic process
negative regulation of DNA-binding transcription factor activity
positive regulation of JUN kinase activity
negative regulation of transcription, DNA-templated
positive regulation of JNK cascade
protein autophosphorylation
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
JUN kinase kinase kinase activity
ATP binding
protein homodimerization activity
bHLH transcription factor binding
apoptotic process
signal transduction
smoothened signaling pathway
JNK cascade
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of apoptotic process
negative regulation of DNA-binding transcription factor activity
positive regulation of JUN kinase activity
negative regulation of transcription, DNA-templated
positive regulation of JNK cascade
protein autophosphorylation
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
JUN kinase kinase kinase activity
ATP binding
protein homodimerization activity
bHLH transcription factor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19007612.hg.1 | ENST00000253055.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19007612.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19007612.hg.1 | TC19000559.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | TC1900008090.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161042.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | NM_002446.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | TC1900008090.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | ENST00000253055.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | ENST00000593906.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | ENST00000597986.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900161043.hg.1 | NM_002446.3 | Refseq |
