Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030815.hg.1 |
ENST00000234310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030815.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030815.hg.1 |
ENST00000409752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030815.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030815.hg.1 |
TC02005028.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030817.hg.1 |
ENST00000234310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030817.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030817.hg.1 |
TC02005028.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200226585.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226585.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226585.hg.1 |
TC0200016682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200226585.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226585.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
TC0200016682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226582.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000234310.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
ENST00000409752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016682.hg.1 |
NM_000945.3 |
Refseq |