Assay Details
Gene Symbol: GADD45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015675.3 | NP_056490.2 | 1 - 2 | 281 | 73 | |
XM_017026822.1 | XP_016882311.1 | 1 - 2 | 279 | 73 | ||
GenBank mRNA | AF078077.1 | - | 1 - 2 | 147 | 73 | |
AF087853.1 | - | 1 - 2 | 268 | 73 | ||
AF090950.1 | - | 1 - 2 | 76 | 73 | ||
AK293087.1 | - | 1 - 2 | 279 | 73 | ||
BC024654.3 | - | 1 - 2 | 187 | 73 | ||
BC113466.1 | - | 1 - 2 | 138 | 8322518 | 73 | |
BC117469.1 | - | 1 - 2 | 138 | 40126020 | 73 | |
BP225713.1 | - | 1 - 2 | 283 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000055.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
apoptotic process
response to stress
multicellular organism development
cell differentiation
positive regulation of apoptotic process
positive regulation of JNK cascade
regulation of cell cycle
positive regulation of p38MAPK cascade
protein binding
negative regulation of protein kinase activity
apoptotic process
response to stress
multicellular organism development
cell differentiation
positive regulation of apoptotic process
positive regulation of JNK cascade
regulation of cell cycle
positive regulation of p38MAPK cascade
protein binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19000872.hg.1 | ENST00000215631.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19000872.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000872.hg.1 | TC19000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000868.hg.1 | ENST00000215631.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19000868.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000868.hg.1 | TC19000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19000870.hg.1 | ENST00000215631.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19000870.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000870.hg.1 | TC19000055.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19000869.hg.1 | ENST00000215631.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19000869.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19000869.hg.1 | TC19000055.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | TC1900006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145955.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | TC1900006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145962.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006588.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | TC1900006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145954.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | NM_015675.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | TC1900006588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000215631.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000585359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000586759.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000587345.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000587887.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000592937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | ENST00000593043.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900145958.hg.1 | NM_015675.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 106600 | - | - |
