Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159531.1 | NP_001153003.1 | 3 - 4 | 384 | 76 | |
NM_020836.3 | NP_065887.1 | 4 - 5 | 310 | 76 | ||
XM_005267919.3 | XP_005267976.1 | 4 - 5 | 327 | 76 | ||
XM_005267920.4 | XP_005267977.1 | 4 - 5 | 327 | 76 | ||
XM_005267921.3 | XP_005267978.1 | 4 - 5 | 4018 | 76 | ||
XM_011537031.1 | XP_011535333.1 | 4 - 5 | 409 | 76 | ||
XM_017021521.1 | XP_016877010.1 | 4 - 5 | 526 | 76 | ||
XM_017021522.1 | XP_016877011.1 | 4 - 5 | 298 | 76 | ||
GenBank mRNA | AB040879.1 | - | 2 - 3 | 3446 | 76 | |
BC002607.2 | - | 4 - 5 | 310 | 3161373 | 76 | |
BG333264.1 | - | 3 - 4 | 385 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001510.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554274.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554274.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000554747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14016955.hg.1 | ENST00000355173.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14016955.hg.1 | ENST00000443071.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14016955.hg.1 | ENST00000556751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14016955.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14016955.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14016955.hg.1 | TC14001510.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000554747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010198.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | NM_020836.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | TC1400010198.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000355173.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000443071.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000553553.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000554140.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000554356.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000556188.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000556751.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | ENST00000557378.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | NM_001159531.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400175534.hg.1 | NM_020836.3 | Refseq |
