Assay Details
Gene Symbol: FN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306129.1 | NP_001293058.1 | 21 - 22 | 3613 | 58 | |
NM_001306130.1 | NP_001293059.1 | 21 - 22 | 3613 | 58 | ||
NM_001306131.1 | NP_001293060.1 | 21 - 22 | 3613 | 58 | ||
NM_001306132.1 | NP_001293061.1 | 21 - 22 | 3613 | 58 | ||
NM_002026.3 | NP_002017.1 | 21 - 22 | 3611 | 58 | ||
NM_212474.2 | NP_997639.1 | 21 - 22 | 3611 | 58 | ||
NM_212476.2 | NP_997641.1 | 21 - 22 | 3611 | 58 | ||
NM_212478.2 | NP_997643.1 | 21 - 22 | 3611 | 58 | ||
NM_212482.2 | NP_997647.1 | 21 - 22 | 3611 | 58 | ||
XM_005246397.1 | XP_005246454.1 | 21 - 22 | 3710 | 58 | ||
XM_005246398.1 | XP_005246455.1 | 21 - 22 | 3710 | 58 | ||
XM_005246399.1 | XP_005246456.1 | 21 - 22 | 3710 | 58 | ||
XM_005246401.1 | XP_005246458.1 | 21 - 22 | 3710 | 58 | ||
XM_005246402.1 | XP_005246459.1 | 21 - 22 | 3710 | 58 | ||
XM_005246403.1 | XP_005246460.1 | 21 - 22 | 3710 | 58 | ||
XM_005246404.1 | XP_005246461.1 | 21 - 22 | 3710 | 58 | ||
XM_005246405.1 | XP_005246462.1 | 21 - 22 | 3710 | 58 | ||
XM_005246406.1 | XP_005246463.1 | 21 - 22 | 3710 | 58 | ||
XM_005246407.1 | XP_005246464.1 | 21 - 22 | 3710 | 58 | ||
XM_005246408.1 | XP_005246465.1 | 21 - 22 | 3710 | 58 | ||
XM_005246409.1 | XP_005246466.1 | 21 - 22 | 3710 | 58 | ||
XM_005246410.1 | XP_005246467.1 | 21 - 22 | 3710 | 58 | ||
XM_005246411.1 | XP_005246468.1 | 21 - 22 | 3710 | 58 | ||
XM_005246412.1 | XP_005246469.1 | 21 - 22 | 3710 | 58 | ||
XM_005246414.1 | XP_005246471.1 | 21 - 22 | 3710 | 58 | ||
XM_005246416.1 | XP_005246473.1 | 21 - 22 | 3710 | 58 | ||
XM_017003692.1 | XP_016859181.1 | 21 - 22 | 3710 | 58 | ||
XM_017003693.1 | XP_016859182.1 | 21 - 22 | 3710 | 58 | ||
XM_017003694.1 | XP_016859183.1 | 21 - 22 | 3710 | 58 | ||
XM_017003695.1 | XP_016859184.1 | 21 - 22 | 3710 | 58 | ||
GenBank mRNA | AB191261.1 | - | 21 - 22 | 3611 | 58 | |
AB209840.1 | - | 21 - 22 | 3603 | 58 | ||
AL832202.1 | - | 21 - 22 | 3612 | 58 | ||
AL832771.1 | - | 21 - 22 | 3612 | 58 | ||
BC117176.1 | - | 21 - 22 | 3591 | 40125727 | 58 | |
BC143754.1 | - | 21 - 22 | 3591 | 9052268 | 58 | |
BC143763.1 | - | 21 - 22 | 3591 | 9052277 | 58 | |
BX537590.1 | - | 21 - 22 | 3612 | 58 | ||
BX538017.1 | - | 21 - 22 | 3612 | 58 | ||
BX538018.1 | - | 21 - 22 | 3612 | 58 | ||
BX640608.1 | - | 21 - 22 | 3611 | 58 | ||
BX640731.1 | - | 21 - 22 | 3612 | 58 | ||
BX640802.1 | - | 21 - 22 | 3613 | 58 | ||
BX640803.1 | - | 21 - 22 | 3611 | 58 | ||
BX640875.1 | - | 21 - 22 | 3612 | 58 | ||
BX640920.1 | - | 1 - 2 | 47 | 58 | ||
BX640999.1 | - | 16 - 17 | 2587 | 58 | ||
BX649182.1 | - | 21 - 22 | 3613 | 58 | ||
CR749281.1 | - | 21 - 22 | 3650 | 58 | ||
CR749316.1 | - | 21 - 22 | 3610 | 58 | ||
CR749317.1 | - | 21 - 22 | 3610 | 58 | ||
CR749666.1 | - | 1 - 2 | 226 | 58 | ||
X02761.1 | - | 21 - 22 | 3265 | 58 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002747.hg.1
Additional Information:
This assay is in the 5'UTR region of the CR749666.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
