Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018930.hg.1 |
ENST00000269554.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018930.hg.1 |
ENST00000311500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018930.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018930.hg.1 |
TC17001433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
TC1700010545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183001.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
TC1700010545.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183000.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000613767.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000617781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
ENST00000619039.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010545.hg.1 |
NM_003559.4 |
Refseq |