Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012211.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030750.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030750.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030750.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030750.hg.1 |
TC01001985.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030748.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030748.hg.1 |
ENST00000366538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030748.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030748.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030748.hg.1 |
TC01001985.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030745.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030745.hg.1 |
TC01001985.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030753.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030753.hg.1 |
ENST00000366538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030753.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030753.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030753.hg.1 |
TC01001985.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030749.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030749.hg.1 |
ENST00000366538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030749.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030749.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030749.hg.1 |
TC01001985.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
TC0100012211.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189899.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189897.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189897.hg.1 |
TC0100012211.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189897.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189893.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189893.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189893.hg.1 |
TC0100012211.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189893.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189893.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
ENST00000358704.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
ENST00000622512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_001278196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_006352.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_205768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012211.hg.1 |
NM_205768.2 |
Refseq |