Assay Details
Gene Symbol: MAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005911.5 | NP_005902.1 | 1 - 2 | 395 | 76 | |
GenBank mRNA | AK291126.1 | - | 1 - 2 | 203 | 76 | |
AK293841.1 | - | 1 - 2 | 205 | 76 | ||
BC001686.2 | - | 1 - 2 | 192 | 3010820 | 76 | |
BC001854.2 | - | 1 - 2 | 192 | 3010820 | 76 | |
BI549760.1 | - | 1 - 2 | 214 | 76 | ||
DC322518.1 | - | 1 - 2 | 236 | 76 | ||
DQ083239.1 | - | 1 - 2 | 131 | 76 | ||
X68836.1 | - | 1 - 2 | 150 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000511.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02007852.hg.1 | TC02000511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007852.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02007855.hg.1 | TC02000511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007855.hg.1 | ENST00000306434.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007855.hg.1 | ENST00000424323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007855.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02007856.hg.1 | TC02000511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007856.hg.1 | ENST00000306434.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007856.hg.1 | ENST00000424323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007856.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02007863.hg.1 | TC02000511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007863.hg.1 | ENST00000306434.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007863.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007863.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008438.hg.1 | ENST00000551090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007857.hg.1 | TC02000511.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007857.hg.1 | ENST00000306434.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007857.hg.1 | ENST00000424323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007857.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157198.hg.1 | TC0200008259.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | ENST00000490878.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008259.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157197.hg.1 | TC0200008259.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000306434.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000409017.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000465151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000469221.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | ENST00000481412.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157202.hg.1 | TC0200008259.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200008438.hg.1 | ENST00000551090.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157194.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157194.hg.1 | NM_005911.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157194.hg.1 | TC0200008259.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 158350 | - | - |
