Assay Details
Gene Symbol: MCRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK054811.1 | - | 1 - 2 | 1488 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000307650.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000474840.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000491999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000575894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000615744.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000619674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000620462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | ENST00000629534.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011322.hg.1 | NM_138418.3 | Refseq |
