Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000369085.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
ENST00000450186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009090.hg.1 |
NM_004281.3 |
Refseq |