Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030560.hg.1 |
ENST00000366557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030560.hg.1 |
ENST00000366558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030560.hg.1 |
ENST00000366559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030560.hg.1 |
NM_003679.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030560.hg.1 |
TC01001974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030561.hg.1 |
ENST00000366557.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030561.hg.1 |
ENST00000366558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030561.hg.1 |
ENST00000366559.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030561.hg.1 |
NM_003679.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030561.hg.1 |
TC01001974.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000366557.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000366558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000366559.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000431245.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
NM_003679.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
TC0100012167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000431245.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000366557.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000366558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
ENST00000366559.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189603.hg.1 |
NM_003679.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000366557.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000366558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000366559.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000431245.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
NM_003679.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
TC0100012167.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000366557.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000366558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000366559.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
ENST00000431245.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189604.hg.1 |
NM_003679.4 |
Refseq |