Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053491.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053491.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053491.hg.1 |
TC01003416.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000468311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000468311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
ENST00000468311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222980.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222980.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222980.hg.1 |
TC0100016158.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100222980.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222980.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000368010.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000468311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000468311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
ENST00000468311.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016158.hg.1 |
NM_012394.3 |
Refseq |