Assay Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020945.1 | NP_065996.1 | 27 - 28 | 4819 | 79 | |
XM_005270004.3 | XP_005270061.1 | 27 - 28 | 4934 | 79 | ||
XM_011539986.2 | XP_011538288.2 | 27 - 28 | 4937 | 79 | ||
XM_011539987.2 | XP_011538289.2 | 27 - 28 | 4940 | 79 | ||
XM_011539988.2 | XP_011538290.1 | 28 - 29 | 5018 | 79 | ||
XM_011539990.2 | XP_011538292.1 | 27 - 28 | 4965 | 79 | ||
XM_011539991.2 | XP_011538293.1 | 27 - 28 | 4969 | 79 | ||
XM_011539992.2 | XP_011538294.1 | 27 - 28 | 4969 | 79 | ||
XM_017016463.1 | XP_016871952.1 | 28 - 29 | 4861 | 79 | ||
XM_017016464.1 | XP_016871953.1 | 9 - 10 | 1248 | 79 | ||
XM_017016465.1 | XP_016871954.1 | 27 - 28 | 4946 | 79 | ||
GenBank mRNA | AK074085.1 | - | 7 - 8 | 877 | 79 | |
AK093607.1 | - | 4 - 5 | 381 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000323.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
