Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | M13929.1 | - | 1 - 2 | 318 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000259523.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000377970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000517291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000520751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000524013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000613283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | ENST00000621592.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008845.hg.1 | NM_002467.5 | Refseq |
