Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000439765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
ENST00000444532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008022.hg.1 |
NM_001243537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006841.hg.1 |
ENST00000512987.1 |
Ensembl |