Assay Details
Gene Symbol: TP53TG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB007456.1 | - | 1 - 2 | 212 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700008250.hg.1 | ENST00000612852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008250.hg.1 | ENST00000612852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000359941.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000416560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000421293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000432193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000542586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000610086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | ENST00000616845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011703.hg.1 | NR_015381.1 | Refseq |
