Assay Details
Gene Symbol: FNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032532.2 | NP_115921.2 | 20 - 21 | 5485 | 95 | |
XM_011536190.2 | XP_011534492.1 | 19 - 20 | 5408 | 95 | ||
XM_011536191.2 | XP_011534493.1 | 17 - 18 | 5126 | 95 | ||
GenBank mRNA | AB058769.2 | - | 18 - 19 | 4953 | 95 | |
AJ586132.1 | - | 10 - 11 | 1386 | 95 | ||
AL832410.1 | - | 10 - 11 | 1684 | 95 | ||
BC146783.1 | - | 18 - 19 | 4953 | 9007239 | 95 | |
BC150607.1 | - | 19 - 20 | 5160 | 9056977 | 95 | |
DQ009660.1 | - | 20 - 21 | 5467 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001142.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
