Assay Details
Gene Symbol: FNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032532.2 | NP_115921.2 | 7 - 8 | 1086 | 95 | |
XM_011536190.2 | XP_011534492.1 | 6 - 7 | 1009 | 95 | ||
XM_011536191.2 | XP_011534493.1 | 4 - 5 | 727 | 95 | ||
GenBank mRNA | AB058769.2 | - | 6 - 7 | 761 | 95 | |
BC146783.1 | - | 6 - 7 | 761 | 9007239 | 95 | |
BC150607.1 | - | 6 - 7 | 761 | 9056977 | 95 | |
DQ009660.1 | - | 7 - 8 | 1086 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001142.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06013580.hg.1 | ENST00000297267.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06013580.hg.1 | ENST00000340366.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06013580.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06013580.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06013580.hg.1 | TC06001142.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | TC0600010040.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600168575.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000297267.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000329629.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | ENST00000480856.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010040.hg.1 | NM_032532.2 | Refseq |
