Assay Details
Gene Symbol: VANGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172411.1 | NP_001165882.1 | 6 - 7 | 1343 | 75 | |
NM_001172412.1 | NP_001165883.1 | 6 - 7 | 1293 | 75 | ||
NM_138959.2 | NP_620409.1 | 6 - 7 | 1349 | 75 | ||
GenBank mRNA | AB057596.1 | - | 6 - 7 | 1293 | 75 | |
AB075805.1 | - | 5 - 6 | 1090 | 75 | ||
AF481859.1 | - | 6 - 7 | 1305 | 75 | ||
BC032773.1 | - | 4 - 5 | 1017 | 75 | ||
BC065272.1 | - | 6 - 7 | 1326 | 6165098 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001012.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | TC01001012.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | ENST00000310260.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | ENST00000355485.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | ENST00000369510.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01016110.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000474344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000474344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000474344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000478369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000478369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | ENST00000478369.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100168564.hg.1 | TC0100009524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000310260.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000355485.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000369510.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000474344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000474344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000474344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000478369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000478369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | ENST00000478369.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172411.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_001172412.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009524.hg.1 | NM_138959.2 | Refseq |
