Assay Details
Gene Symbol: HTR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006028.4 | NP_006019.1 | 1 - 2 | 186 | 84 | |
GenBank mRNA | AF080582.1 | - | 1 - 2 | 115 | 84 | |
AF169255.1 | - | 1 - 2 | 189 | 84 | ||
AK314268.1 | - | 1 - 2 | 186 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001024.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
serotonin receptor signaling pathway
synaptic transmission, cholinergic
ion transmembrane transport
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
G-protein coupled serotonin receptor activity
serotonin-activated cation-selective channel activity
acetylcholine receptor activity
acetylcholine binding
serotonin receptor signaling pathway
synaptic transmission, cholinergic
ion transmembrane transport
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
G-protein coupled serotonin receptor activity
serotonin-activated cation-selective channel activity
acetylcholine receptor activity
acetylcholine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11012795.hg.1 | ENST00000260191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012795.hg.1 | ENST00000537778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012795.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012795.hg.1 | TC11001024.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11012792.hg.1 | ENST00000260191.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012792.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012792.hg.1 | TC11001024.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167434.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167434.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167434.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167434.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167434.hg.1 | TC1100009097.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100167433.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167433.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167433.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167433.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167433.hg.1 | TC1100009097.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | ENST00000543092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009097.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | ENST00000260191.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | ENST00000537778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | NM_006028.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167436.hg.1 | TC1100009097.hg.1 | Clariom D TC |
