Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035972.hg.1 |
ENST00000290167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035972.hg.1 |
ENST00000542383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035972.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035972.hg.1 |
TC01002330.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100198252.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198252.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198252.hg.1 |
TC0100013246.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198252.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198252.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000290167.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000415567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
ENST00000441048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013246.hg.1 |
NM_030761.4 |
Refseq |