Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015403.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01015403.hg.1 |
TC01000967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000420853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000420853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000420853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000462836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000462836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000462836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167512.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100167512.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167512.hg.1 |
TC0100009387.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100167512.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100167512.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000420853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000420853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000420853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000462836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000462836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000462836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
ENST00000474680.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009387.hg.1 |
NR_003599.2 |
Refseq |