Assay Details
Gene Symbol: GLDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000170.2 | NP_000161.2 | 6 - 7 | 1057 | 153 | |
GenBank mRNA | AK310534.1 | - | 2 - 3 | 432 | 153 | |
AK314156.1 | - | 6 - 7 | 1059 | 153 | ||
BC111993.1 | - | 6 - 7 | 930 | 8327461 | 153 | |
BC111995.1 | - | 6 - 7 | 930 | 8327463 | 153 | |
D90239.1 | - | 6 - 7 | 1014 | 153 | ||
M63635.1 | - | 6 - 7 | 945 | 153 | ||
M64590.1 | - | 6 - 7 | 1014 | 153 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000899.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09011954.hg.1 | TC09000899.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09011954.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09011954.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900165105.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900165105.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900165105.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900165105.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900165105.hg.1 | TC0900009489.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
