Assay Details
Gene Symbol: PIP4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003559.4 | NP_003550.1 | 4 - 5 | 981 | 158 | |
XM_017025196.1 | XP_016880685.1 | 5 - 6 | 558 | 158 | ||
XM_017025198.1 | XP_016880687.1 | 3 - 4 | 944 | 158 | ||
GenBank mRNA | BC027459.1 | - | 4 - 5 | 598 | 3877995 | 158 |
BT019614.1 | - | 4 - 5 | 500 | 158 | ||
U85245.1 | - | 4 - 5 | 980 | 158 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001433.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017025198.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
cell surface receptor signaling pathway
regulation of autophagy
regulation of phosphatidylinositol 3-kinase signaling
phosphatidylinositol phosphate biosynthetic process
positive regulation of autophagosome assembly
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
1-phosphatidylinositol-5-phosphate 4-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol biosynthetic process
cell surface receptor signaling pathway
regulation of autophagy
regulation of phosphatidylinositol 3-kinase signaling
phosphatidylinositol phosphate biosynthetic process
positive regulation of autophagosome assembly
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
1-phosphatidylinositol-5-phosphate 4-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17018929.hg.1 | ENST00000269554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17018929.hg.1 | ENST00000311500.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17018929.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018929.hg.1 | TC17001433.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | TC1700010545.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700182998.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000613767.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000617781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | ENST00000619039.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010545.hg.1 | NM_003559.4 | Refseq |
