Assay Details
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320340.1 | NP_001307269.1 | 2 - 3 | 334 | 124 | |
GenBank mRNA | BI826588.1 | - | 2 - 3 | 333 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of autophagy
negative regulation of protein complex assembly
positive regulation of protein lipidation
negative regulation of ER-associated ubiquitin-dependent protein catabolic process
negative regulation of retrograde protein transport, ER to cytosol
negative regulation of VCP-NPL4-UFD1 AAA ATPase complex assembly
protein binding
protein self-association
ATPase binding
positive regulation of autophagy
negative regulation of protein complex assembly
positive regulation of protein lipidation
negative regulation of ER-associated ubiquitin-dependent protein catabolic process
negative regulation of retrograde protein transport, ER to cytosol
negative regulation of VCP-NPL4-UFD1 AAA ATPase complex assembly
protein binding
protein self-association
ATPase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000529848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000529848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000529848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000354193.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000525670.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000529848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000529848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000529848.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000530199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | ENST00000533774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010349.hg.1 | NM_148893.2 | Refseq |
