Assay Details
Gene Symbol: TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291333.1 | NP_001278262.1 | 6 - 7 | 763 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | TC0100008402.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159850.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | TC0100008402.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159849.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | TC0100008402.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159848.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000486091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NR_037641.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | TC0100008402.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000371281.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000371284.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000414150.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000425300.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000460155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000486091.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | ENST00000606515.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NM_001291333.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NM_004623.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NR_037639.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NR_037640.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100159856.hg.1 | NR_037641.2 | Refseq |
