Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015603.hg.1 |
TC08001209.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015603.hg.1 |
ENST00000358543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015603.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170816.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170816.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170816.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170816.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170816.hg.1 |
TC0800010426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000358543.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
ENST00000523939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010426.hg.1 |
NM_207413.3 |
Refseq |