Assay Details
Gene Symbol: IFNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 49 | 157 | |
GenBank mRNA | BC074991.2 | - | 1 | 47 | 30915521 | 157 |
BC074992.2 | - | 1 | 47 | 30915307 | 157 | |
BC113994.1 | - | 1 | 49 | 40084821 | 157 | |
BC114000.1 | - | 1 | 49 | 40084820 | 157 | |
M34913.1 | - | 1 | 49 | 157 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09002913.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC074991.2,BC074992.2,BC113994.1,BC114000.1,M34913.1,NM_021057.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
cell-cell signaling
blood coagulation
response to virus
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
innate immune response
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
cytokine activity
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
cell-cell signaling
blood coagulation
response to virus
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
innate immune response
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
cytokine activity
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16164853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16164847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925748 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06637766 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06637767 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16164849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925772 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 530721 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 407411 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345086 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12189 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36089 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02075404 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3241224 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345086 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3398932 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3345086 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637768 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925772 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164853 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925772 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637768 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164853 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637767 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06637768 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06637767 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | AB058771.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | AK022646.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | AY219922.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | AY929062.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | BC004514.2 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | BC065271.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | BX537956.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | CR608697.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 141649 | NM_025139.3 | refseq_mrna_human_NM |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32403125 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06637766 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16164847 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06637771 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06637767 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_021057_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160386 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160380 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160398 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160379 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160385 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160396 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8160394 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8160383 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8160377 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208259_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738439_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738439_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR09012687.hg.1 | TC09002913.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09012687.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09012687.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09012684.hg.1 | TC09002913.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09012684.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09012684.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166332.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166332.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166332.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166332.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166332.hg.1 | TC0900009696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900166329.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166329.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166329.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166329.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166329.hg.1 | TC0900009696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900166333.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166333.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166333.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166333.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166333.hg.1 | TC0900009696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009696.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166331.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166331.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166331.hg.1 | ENST00000239347.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900166331.hg.1 | NM_021057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900166331.hg.1 | TC0900009696.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 208259_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208259_x_at | - | - |
Human Genome U133A 2.0 Array | 208259_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 208259_x_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_021057_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw526816 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208259_x_at3 | - | - |
Human Genome U133 Set | 208259_PM_x_at | - | - |
Human Genome U133 Set | 208259_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037176 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037176 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 118924 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1776023 | BC113994.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1776023 | BC114000.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1776023 | M34913.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2186745 | BC074991.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2186745 | BC074992.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2186745 | BC113994.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776023 | BC113994.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776023 | BC114000.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776023 | M34913.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1776023 | BC113994.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1776023 | BC114000.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1776023 | M34913.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2186745 | BC074991.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2186745 | BC074992.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2186745 | BC113994.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2186745 | BC074991.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2186745 | BC074992.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2186745 | BC113994.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2186745 | BC074991.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2186745 | BC074992.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2186745 | BC113994.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_10800141-S | M34913.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_27442-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0000110075 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0000110075 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1776023 | BC113994.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1776023 | BC114000.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1776023 | M34913.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776023 | BC113994.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776023 | BC114000.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776023 | M34913.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 3060500 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27442 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2186745 | BC074991.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2186745 | BC074992.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2186745 | BC113994.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776023 | BC113994.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776023 | BC114000.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776023 | M34913.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2186745 | BC074991.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2186745 | BC074992.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2186745 | BC113994.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2186745 | NM_021057.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2186745 | BC074991.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2186745 | BC074992.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2186745 | BC113994.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2186745 | BC114000.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776023 | NM_021057.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776023 | BC113994.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776023 | BC114000.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776023 | M34913.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1776023 | NM_021057.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1776023 | BC113994.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1776023 | BC114000.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1776023 | M34913.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2186745 | NM_021057.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2186745 | BC074991.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2186745 | BC074992.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2186745 | BC113994.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2186745 | BC114000.1 | human_transcriptome |
