Assay Details
Gene Symbol: LL22NC03-75H12.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109967.1 | 1 | 217 | 102 | ||
GenBank mRNA | BC037972.1 | - | 1 | 217 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
